trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl ≥97%

Supplier: BeanTown Chemical

Synonyms: trans,trans-4-(3,4-Difluorophenyl)-4'-pentyl-1,1'-bi(cyclohexane), trans,trans-4-(3,4-Difluorophenyl)-4'-pentylbicyclohexyl

133505-1G 133505-5G
BT133505-1GEA 103 USD
BT133505-1G BT133505-5G
trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl ≥97%
trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl

CAS: 118164-51-5; MDL No: MFCD13182310 Solid; Molecular Formula: C23H34F2; MW: 348.52 Melting Point: 45-47°


  • Solid

Formula: C₂₃H₃₄F₂
MW: 348.522 g/mol
Melting Pt: 45…47 °C
MDL Number: MFCD13182310
CAS Number: 118164-51-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR