Ruthenium(III) nitrosylacetate ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

133495-1G 133495-5G
BT133495-1GEA 117 USD
BT133495-1G BT133495-5G
Ruthenium(III) nitrosylacetate ≥99.99% (trace metals basis)
Ruthenium(III) nitrosylacetate

MDL No: MFCD02684507 Powder; Linear Formula: Ru(NO)(OOCCH3)3; MW: 308.18


  • Powder

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR