Iron(II) sulfide ≥99.95% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Ferrous sulphide
133470-25G
133470-5G
BT133470-25GEA
302
USD
BT133470-25G
BT133470-5G
Iron(II) sulfide ≥99.95% (trace metals basis)
Iron(II) sulphide
CAS: 1317-37-9; EC No: 215-268-6; MDL No: MFCD00011013
Powder; Molecular Formula: FeS; MW: 87.91
Density (g/mL): 4.74
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Powder
Formula:
FeS MW: 87.91 g/mol Density: 4.74 g/mL |
MDL Number:
MFCD00011013 CAS Number: 1317-37-9 EINECS: 215-268-6 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99.95% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...