(S)-(+)-4-Phenyl-2-oxazolidinone ≥98%

Supplier: BeanTown Chemical

Synonyms: (S)-4-Phenyloxazolidin-2-one

133445-1G 133445-5G
BT133445-1GEA 92 USD
BT133445-1G BT133445-5G
(S)-(+)-4-Phenyl-2-oxazolidinone ≥98%
(S)-(+)-4-Phenyl-2-oxazolidinone

CAS: 99395-88-7; MDL No: MFCD00043396 Crystalline; Molecular Formula: C9H9NO2 ; MW: 163.17 Melting Point: 129-132° Optical Rotation: [α]20/D +48°, c = 2 in chloroform


  • Optical rotation: [α]20/D +48°, c = 2 in chloroform
  • Crystalline

Formula: C₉H₉NO₂
MW: 163.18 g/mol
Melting Pt: 129…132 °C
MDL Number: MFCD00043396
CAS Number: 99395-88-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR