Ethyl-6-oxo-2-(trifluoromethyl)-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%

Supplier: BeanTown Chemical

Synonyms: Ethyl 6-oxo-1,4,5,6-tetrahydro-2-(trifluoromethyl)pyridine-3-carboxylate, Ethyl 6-oxo-2-(trifluoromethyl)-1,4,5,6-tetrahydropyridine-3-carboxylate

133420-1G 133420-5G
BT133420-1GEA 139 USD
BT133420-1G BT133420-5G
Ethyl-6-oxo-2-(trifluoromethyl)-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%
Ethyl-6-oxo-2-(trifluoromethyl)-1,4,5,6-tetrahydro-3-pyridinecarboxylate

CAS: 194673-12-6; EC No: 260-171-4; MDL No: MFCD00975005 Solid; Molecular Formula: C9H10F3NO3; MW: 237.18 Melting Point: 128-130° Density (g/mL): 1.351


  • Solid

Formula: C₉H₁₀F₃NO₃
MW: 237.178 g/mol
Melting Pt: 128…130 °C
Density: 1.351 g/mL
MDL Number: MFCD00975005
CAS Number: 194673-12-6
EINECS: 260-171-4

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR