Bis(tri-tert-butylphosphine)palladium(0)
Supplier: BeanTown Chemical
133390-1G
133390-250MG
BT133390-1GEA
322
USD
BT133390-1G
BT133390-250MG
Bis(tri-tert-butylphosphine)palladium(0)
Bis(tri-tert-butylphosphine)palladium(0)
CAS: 53199-31-8; MDL No: MFCD03094580
Powder, packaged under argon; Molecular Formula: C24H54P2Pd ; MW: 511.05
Melting Point: 258-272°
Air Sensitive
- Air Sensitive
- Powder, packaged under argon
Formula:
C₂₄H₅₄P₂Pd Melting Pt: 258…272 °C |
MDL Number:
MFCD03094580 CAS Number: 53199-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...