2-Bromo-5-methylpyridine ≥98%

Supplier: BeanTown Chemical

133375-100G 133375-25G 133375-5G
BT133375-100GEA 477.58 USD
BT133375-100G BT133375-25G BT133375-5G
2-Bromo-5-methylpyridine ≥98%
2-Bromo-5-methylpyridine

CAS: 3510-66-5; MDL No: MFCD00209553 Solid; Molecular Formula: C6H6BrN ; MW: 172.02 Melting Point: 41-43°; Boiling Point: 95-96°/12.5 mmHg; Flash point: 103°C (217°F)


  • Solid

Formula: C₆H₆BrN
Boiling Pt: 95…96 °C/12.5 mmHg
Melting Pt: 41…43 °C
Flash Pt: 103 °C (217 °F)
MDL Number: MFCD00209553
CAS Number: 3510-66-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR