L-Valine methyl ester hydrochloride ≥99%

Supplier: BeanTown Chemical

Synonyms: H-Val-OMe.HCl, L-2-Aminoisovaleric Acid Methyl Ester Hydrochloride, L-Methyl valinate hydrochloride, H-Val-OMe·HCl

133360-10G 133360-50G
BT133360-10GEA 53 USD
BT133360-10G BT133360-50G
L-Valine methyl ester hydrochloride ≥99%
L-Valine methyl ester hydrochloride

CAS: 6306-52-1; EC No: 228-620-9; MDL No: MFCD00012497 Crystalline; Molecular Formula: C6H13NO2·HCl; MW: 167.63 Melting Point: 171-173° Optical Rotation: [α]/D +15°, c = 2 in water


  • Optical rotation: [α]/D +15°, c = 2 in water
  • Crystalline

Formula: C₆H₁₄ClNO₂
MW: 167.63 g/mol
Melting Pt: 171…173 °C
MDL Number: MFCD00012497
CAS Number: 6306-52-1
EINECS: 228-620-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR