Sodium trifluoromethanesulfonate
Supplier: BeanTown Chemical
Synonyms:
Trifluoromethanesulphonic acid sodium salt
133345-25G
133345-5G
BT133345-25GEA
182.02
USD
BT133345-25G
BT133345-5G
Sodium trifluoromethanesulfonate
Sodium trifluoromethanesulphonate
CAS: 2926-30-9; MDL No: MFCD00061607
Crystalline; Linear Formula: CF3SO3Na; MW: 172.06
Melting Point: 253-255°
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
CF₃NaO₃S MW: 172.06 g/mol Melting Pt: 253…255 °C |
MDL Number:
MFCD00061607 CAS Number: 2926-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...