1H-Pyrazole-3-carboxylic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
3-Pyrazolecarboxylic acid, Pyrazole-3-carboxylic acid
133285-1G
133285-5G
BT133285-1GEA
65
USD
BT133285-1G
BT133285-5G
1H-Pyrazole-3-carboxylic acid ≥97%
1H-Pyrazole-3-carboxylic acid
CAS: 1621-91-6; MDL No: MFCD00077436
Solid; Molecular Formula: C4H4N2O2; MW: 112.09
Melting Point: 204-213°
- Solid
Formula:
C₄H₄N₂O₂ MW: 112.09 g/mol Melting Pt: 204…213 °C |
MDL Number:
MFCD00077436 CAS Number: 1621-91-6 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...