DL-α-Tocopherol acetate ≥97%

Supplier: BeanTown Chemical

133255-100G 133255-25G 133255-500G
BT133255-100GEA 77.41 USD
BT133255-100G BT133255-25G BT133255-500G
DL-α-Tocopherol acetate ≥97%
DL-α-Tocopherol acetate

CAS: 7695-91-2; EC No: 231-710-0; MDL No: MFCD00072042; RTECS: GA8747000 Liquid; Molecular Formula: C31H52O3; MW: 472.75 Melting Point: -28°; Boiling Point: 300°; Flash point: 210°C (410°F) Density (g/mL): 0.953; Refractive Index: 1.496 Air Sensitive, Light Sensitive


  • RTECS: GA8747000
  • Refractive index: 1.496
  • Air Sensitive, Light Sensitive
  • Liquid

Formula: C₃₁H₅₂O₃
Boiling Pt: >300 °C
Melting Pt: –28 °C
Density: 0.953 g/mL
Flash Pt: 210 °C (410 °F)
MDL Number: MFCD00072042
CAS Number: 7695-91-2
EINECS: 231-710-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR