Guanidinium hydrochloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

133240-1KG 133240-250G 133240-5KG
BT133240-1KGEA 72.43 USD
BT133240-1KG BT133240-250G BT133240-5KG
Guanidinium hydrochloride ≥98%
Guanidinium hydrochloride

CAS: 50-01-1; EC No: 200-002-3; MDL No: MFCD00013026; RTECS: MF4300000 Crystalline; Molecular Formula: CH5N3·HCl; MW: 95.53 Melting Point: 180-185° Density (g/mL): 1.344 Hygroscopic


  • RTECS: MF4300000
  • Hygroscopic
  • Crystalline

Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 180…185 °C
Density: 1.344 g/mL
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR