5-Fluorosalicylaldehyde ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Fluoro-2-hydroxybenzaldehyde

133180-1G 133180-5G
BT133180-1GEA 121 USD
BT133180-1G BT133180-5G
5-Fluorosalicylaldehyde ≥98%
5-Fluorosalicylaldehyde

CAS: 347-54-6; MDL No: MFCD01090997 Powder/Flakes; Molecular Formula: C7H5FO2; MW: 140.11 Melting Point: 82-85° Air Sensitive


  • Air Sensitive
  • Powder/Flakes

Formula: C₇H₅FO₂
MW: 140.11 g/mol
Melting Pt: 82…85 °C
MDL Number: MFCD01090997
CAS Number: 347-54-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR