6-Bromoquinolin-4-ol ≥97%

Supplier: BeanTown Chemical

Synonyms: 6-Bromo-4-quinolinol, 6-Bromo-4-hydroxyquinoline

133175-1G 133175-250MG
BT133175-1GEA 419 USD
BT133175-1G BT133175-250MG
6-Bromoquinolin-4-ol ≥97%
6-Bromoquinolin-4-ol

CAS: 145369-94-4; MDL No: MFCD00272435 Solid; Molecular Formula: C9H6BrNO ; MW: 224.05


  • Solid

Formula: C₉H₆BrNO
MDL Number: MFCD00272435
CAS Number: 145369-94-4

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR