4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate ≥98%
Supplier: BeanTown Chemical
Synonyms:
BOC-L-Aspartic acid 4-benzylester, 4-Benzyl N-Boc-L-aspartate, Boc-Asp(Obzl)-OH, Boc-Asp(OBzl)-OH, N-Boc-L-aspartic Acid 4-Benzyl Ester, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Benzyl Ester
133170-1G
133170-5G
BT133170-1GEA
21
USD
BT133170-1G
BT133170-5G
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate ≥98%
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate
CAS: 7536-58-5; EC No: 231-406-8; MDL No: MFCD00065564
Powder; Molecular Formula: C16H21NO6 ; MW: 323.34
Melting Point: 98-102°
Optical Rotation: [α]20/D −20.0±1°, c = 2% in DMF
- Optical rotation: [α]20/D −20.0±1°, c = 2% in DMF
- Powder
Formula:
C₁₆H₂₁NO₆ Melting Pt: 98…102 °C |
MDL Number:
MFCD00065564 CAS Number: 7536-58-5 EINECS: 231-406-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...