Aluminium sulfate hydrate ≥97%

Supplier: BeanTown Chemical

Synonyms: Aluminum(III) sulphate xhydrate

133165-1KG 133165-5KG
BT133165-1KGEA 70.12 USD
BT133165-1KG BT133165-5KG
Aluminium sulfate hydrate ≥97%
Aluminium sulphate hydrate

CAS: 17927-65-0; EC No: 233-135-0; MDL No: MFCD00149136; RTECS: BD1700000 Crystalline; Linear Formula: Al2(SO4)3·xH2O ; MW: 342.15 (anhydrous) Density (g/mL): 1.69


  • RTECS: BD1700000
  • Crystalline

Formula: Al₂(SO₄)₃·xH₂O
MW: 366.21 g/mol
Density: 1.69 g/mL
MDL Number: MFCD00149136
CAS Number: 17927-65-0
EINECS: 233-135-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR