Potassium iodate 0.1 N in water volumetric solution

Supplier: BeanTown Chemical

133140-1L 133140-4L
BT133140-1LEA 52 USD
BT133140-1L BT133140-4L
Potassium iodate 0.1 N in water volumetric solution
Potassium iodate

CAS: 7758-05-6; EC No: 231-831-9; MDL No: MFCD00011406 Liquid; Linear Formula: KIO3; MW: 214.00 Density (g/mL): 1


  • Liquid

Formula: KlO₃
MW: 214 g/mol
Density: 1 g/mL
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR