Holmium(III) oxide ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Diholmium trioxide, Holmium oxide

132945-10G 132945-50G
BT132945-10GEA 170 USD
BT132945-10G BT132945-50G
Holmium(III) oxide ≥99.9%
Holmium(III) oxide

CAS: 12055-62-8; EC No: 235-015-3; MDL No: MFCD00011053 3-12mm Sintered Lump; Molecular Formula: Ho2O3; MW: 377.86 Melting Point: 2330°


  • 3-12mm Sintered Lump

Formula: Ho₂O₃
MW: 377.86 g/mol
Boiling Pt: 2600
Melting Pt: 2400
MDL Number: MFCD00011053
CAS Number: 12055-62-8
EINECS: 235-015-3

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR