6-Methoxy-2-tetralone ≥90%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
6-Methoxy-3,4-dihydronaphthalen-2(1H)-one, 3,4-Dihydro-6-methoxynaphthalen-2(1H)-one
132920-1G
132920-250MG
BT132920-1GEA
165
USD
BT132920-1G
BT132920-250MG
6-Methoxy-2-tetralone ≥90%, Technical Grade
6-Methoxy-2-tetralone
CAS: 2472-22-2; EC No: 219-592-9; MDL No: MFCD00001729
Liquid; Molecular Formula: C11H12O2; MW: 176.21
Melting Point: 30-37°; Flash point:
- Refractive index: 1.565
- Liquid
Formula:
C₁₁H₁₂O₂ MW: 176.22 g/mol Melting Pt: 30…37 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00001729 CAS Number: 2472-22-2 EINECS: 219-592-9 |
Specification Test Results
Assay | 90% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...