3-Fluoro-4-cyanophenyl-4-ethylbenzoate
Supplier: BeanTown Chemical
Synonyms:
4-Cyano-3-fluorophenyl 4-Ethylbenzoate, 4-Ethylbenzoic acid 4-Cyano-3-fluorophenyl ester
132910-1G
132910-5G
BT132910-1GEA
103
USD
BT132910-1G
BT132910-5G
3-Fluoro-4-cyanophenyl-4-ethylbenzoate
3-Fluoro-4-cyanophenyl-4-ethylbenzoate
CAS: 86776-50-3
Solid; Molecular Formula: C16H12FNO2; MW: 269.28
- Solid
Formula:
C₁₆H₁₂FNO₂ |
MDL Number:
MFCD00689047 CAS Number: 86776-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...