Dimethyl sulphoxide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
132780-10KG
132780-2.5KG
132780-500G
BT132780-10KGEA
238.24
USD
BT132780-10KG
BT132780-2.5KG
BT132780-500G
Dimethyl sulphoxide ≥99%
Dimethyl sulphoxide
CAS: 67-68-5; EC No: 200-664-3; MDL No: MFCD00002089; RTECS: PV6210000
Liquid; Linear Formula: (CH3)2SO ; MW: 78.13
Melting Point: 16-19°; Boiling Point: 189°; Flash point: 87°C (189°F)
Density (g/mL): 1.101; Refractive Index: 1.479
Hygroscopic
- RTECS: PV6210000
- Refractive index: 1.479
- Hygroscopic
- Liquid
Formula:
(CH₃)₂SO Boiling Pt: 189 °C Melting Pt: 16…19 °C Density: 1.101 g/mL Flash Pt: 87 °C (189 °F) |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 EINECS: 200-664-3 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...