Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥98%
Supplier: BeanTown Chemical
Synonyms:
Benzo-18-crown 6-Ether
132765-1G
132765-25G
132765-5G
BT132765-1GEA
44
USD
BT132765-1G
BT132765-25G
BT132765-5G
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥98%
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane)
CAS: 14098-24-9; MDL No: MFCD00062741
Granular/Chunks; Molecular Formula: C16H24O6; MW: 312.36
Melting Point: 42-45°; Flash point:
- Hygroscopic
- Granular/Chunks
Formula:
C₁₆H₂₄O₆ MW: 312.36 g/mol Melting Pt: 42…45 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00062741 CAS Number: 14098-24-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...