Sodium-1-decanesulphonate ≥99%

Supplier: BeanTown Chemical

Synonyms: Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt

132740-100G 132740-25G 132740-5G
BT132740-100GEA 536.51 USD
BT132740-100G BT132740-25G BT132740-5G
Sodium-1-decanesulphonate ≥99%
Sodium-1-decanesulphonate

CAS: 13419-61-9; EC No: 236-525-9; MDL No: MFCD00007526 Crystalline; Linear Formula: CH3(CH2)9SO3Na; MW: 244.33 Melting Point: 300° Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₁₀H₂₁NaO₃S
Melting Pt: >300 °C
MDL Number: MFCD00007526
CAS Number: 13419-61-9
EINECS: 236-525-9

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR