Ammonium dihydrogen phosphate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

132695-10KG 132695-2.5KG 132695-500G
BT132695-10KGEA 287 USD
BT132695-10KG BT132695-2.5KG BT132695-500G
Ammonium dihydrogen phosphate ≥98%
Ammonium dihydrogen phosphate

CAS: 7722-76-1; EC No: 231-764-5; MDL No: MFCD00003396 Crystalline; Linear Formula: NH4H2PO4; MW: 115.03 Melting Point: 190° (decomposes)


  • Crystalline

Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190 °C (dec.)
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR