5-Nitroisophthalic acid ≥98%

Supplier: BeanTown Chemical

132690-100G 132690-500G
BT132690-100GEA 28.81 USD
BT132690-100G BT132690-500G
5-Nitroisophthalic acid ≥98%
5-Nitroisophthalic acid

CAS: 618-88-2; EC No: 210-568-3; MDL No: MFCD00007254 Powder; Molecular Formula: C8H3NO6; MW: 211.13 Melting Point: 259-261°; Flash point: 120°C (248°F)


  • Powder

Formula: C₈H₅NO₆
MW: 211.13 g/mol
Melting Pt: 259…261 °C
Flash Pt: 120 °C (248 °F)
MDL Number: MFCD00007254
CAS Number: 618-88-2
EINECS: 210-568-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR