2-Chloro-5-methoxybenzimidazole ≥94%
Supplier: BeanTown Chemical
Synonyms:
2-Chloro-5-methoxy-1H-benzo[d]imidazole, 2-Chloro-5-methoxy-1H-benzimidazole
132685-1G
132685-250MG
BT132685-1GEA
211
USD
BT132685-1G
BT132685-250MG
2-Chloro-5-methoxybenzimidazole ≥94%
2-Chloro-5-methoxybenzimidazole
CAS: 15965-54-5; MDL No: MFCD00051945
Powder; Molecular Formula: C8H7ClN2O; MW: 182.61
- Powder
Formula:
C₈H₇ClN₂O MW: 182.607 g/mol |
MDL Number:
MFCD00051945 CAS Number: 15965-54-5 |
Specification Test Results
Assay | 94% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...