(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: (1R,2R)-Cyclohexane-1,2-dicarboxylic acid, (1R,2R)-(-)-trans-Cyclohexane-1,2-dicarboxylic acid

132675-1G 132675-250MG 132675-5G
BT132675-1GEA 74 USD
BT132675-1G BT132675-250MG BT132675-5G
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid ≥98%
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid

CAS: 46022-05-3; MDL No: MFCD00062973 Solid; Molecular Formula: C8H12O4; MW: 172.18 Melting Point: 184°


  • Solid

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Melting Pt: 184 °C
MDL Number: MFCD00062973
CAS Number: 46022-05-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR