Sodium hydrogen carbonate ≥99%

Supplier: BeanTown Chemical

Synonyms: Sodium bicarbonate

132620-10KG 132620-2.5KG 132620-500G
BT132620-10KGEA 133 USD
BT132620-10KG BT132620-2.5KG BT132620-500G
Sodium hydrogen carbonate ≥99%
Sodium hydrogen carbonate

CAS: 144-55-8; EC No: 205-633-8; MDL No: MFCD00003528; RTECS: VZ0950000 Solid; Linear Formula: NaHCO3; MW: 84.01 Melting Point: 300° Density (g/mL): 2.16; Refractive Index: 1.500


  • RTECS: VZ0950000
  • Refractive index: 1.500
  • Solid

Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 300 °C
Density: 2.16 g/mL
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR