N-(Hydroxymethyl)phthalimide ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-(Hydroxymethyl)isoindoline-1,3-dione

132610-100G 132610-25G 132610-500G
BT132610-100GEA 77.38 USD
BT132610-100G BT132610-25G BT132610-500G
N-(Hydroxymethyl)phthalimide ≥97%
N-(Hydroxymethyl)phthalimide

CAS: 118-29-6; EC No: 204-241-4; MDL No: MFCD00005899; RTECS: TI5270000 Powder; Molecular Formula: C9H7NO3 ; MW: 177.16 Melting Point: 147-149°


  • RTECS: TI5270000
  • Powder

Formula: C₉H₇NO₃
Melting Pt: 147…149 °C
MDL Number: MFCD00005899
CAS Number: 118-29-6
EINECS: 204-241-4

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR