Iron sulfide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Iron pyrite, Iron disulphide

132595-10G 132595-1G 132595-50G
BT132595-10GEA 230.61 USD
BT132595-10G BT132595-1G BT132595-50G
Iron sulfide ≥99.9% (trace metals basis)
Iron sulfide

CAS: 12068-85-8; EC No: 235-106-8; MDL No: MFCD00064690 -100 Mesh Powder; Molecular Formula: FeS2; MW: 119.98 Melting Point: 450°


  • -100 Mesh Powder

Formula: FeS₂
MW: 119.98 g/mol
Melting Pt: 450 °C
MDL Number: MFCD00064690
CAS Number: 12068-85-8
EINECS: 235-106-8

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR