3-Bromo-o-xylene ≥99%

Supplier: BeanTown Chemical

Synonyms: 1-Bromo-2,3-dimethylbenzene

132535-100G 132535-25G 132535-5G
BT132535-100GEA 650 USD
BT132535-100G BT132535-25G BT132535-5G
3-Bromo-o-xylene ≥99%
3-Bromo-o-xylene

CAS: 576-23-8; EC No: 209-398-2; MDL No: MFCD00000069 Liquid; Molecular Formula: C8H9Br ; MW: 185.06 Boiling Point: 214°; Flash point: 80°C (176°F) Density (g/mL): 1.365; Refractive Index: 1.560


  • Refractive index: 1.560
  • Liquid

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 214 °C
Density: 1.365 g/mL
Flash Pt: 80 °C (176 °F)
MDL Number: MFCD00000069
CAS Number: 576-23-8
EINECS: 209-398-2
UN: 1701
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR