Gold(III) oxide ≥99.99%

Supplier: BeanTown Chemical

132520-1G 132520-250MG 132520-5G
BT132520-1GEA 253 USD
BT132520-1G BT132520-250MG BT132520-5G
Gold(III) oxide ≥99.99%
Gold(III) oxide

CAS: 1303-58-8; EC No: 215-122-1; MDL No: MFCD00014173 Powder; Molecular Formula: Au2O3; MW: 441.93


  • Powder

Formula: Au₂O₃
MW: 441.93
Melting Pt: 150
Density: 3.6
MDL Number: MFCD00014173
CAS Number: 1303-58-8
EINECS: 215-122-1
Merck Index: 12,04545

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR