5-Methoxyisatin ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Methoxy-1H-indole-2,3-dione, 5-Methoxyindoline-2,3-dione

132510-100G 132510-25G 132510-5G
BT132510-100GEA 739 USD
BT132510-100G BT132510-25G BT132510-5G
5-Methoxyisatin ≥98%
5-Methoxyisatin

CAS: 39755-95-8; MDL No: MFCD00169023; RTECS: NL7938000 Solid; Molecular Formula: C9H7NO3; MW: 177.16 Melting Point: 199-202°


  • RTECS: NL7938000
  • Solid

Formula: C₉H₇NO₃
MW: 177.16 g/mol
Melting Pt: 199…202 °C
MDL Number: MFCD00169023
CAS Number: 39755-95-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR