4-Nitrophthalimide ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Nitroisoindoline-1,3-dione

132415-100G 132415-25G 132415-5G
BT132415-100GEA 426 USD
BT132415-100G BT132415-25G BT132415-5G
4-Nitrophthalimide ≥98%
4-Nitrophthalimide

CAS: 89-40-7; EC No: 201-905-5; MDL No: MFCD00005884; RTECS: TI5625000 Powder; Molecular Formula: C8H4N2O4 ; MW: 192.13 Melting Point: 195-199°


  • RTECS: TI5625000
  • Powder

Formula: C₈H₄N₂O₄
MW: 192.13 g/mol
Melting Pt: 195…199 °C
MDL Number: MFCD00005884
CAS Number: 89-40-7
EINECS: 201-905-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR