3-Hydroxy-5-methylisoxazole ≥97%

Supplier: BeanTown Chemical

Synonyms: Hymexazol

132395-100G 132395-25G 132395-5G
BT132395-100GEA 452.99 USD
BT132395-100G BT132395-25G BT132395-5G
3-Hydroxy-5-methylisoxazole ≥97%
3-Hydroxy-5-methylisoxazole

CAS: 10004-44-1; EC No: 233-000-6; MDL No: MFCD00144468; RTECS: NY2932000 Powder; Molecular Formula: C4H5NO2; MW: 99.09


  • RTECS: NY2932000
  • Powder

Formula: C₄H₅NO₂
MW: 99.09 g/mol
MDL Number: MFCD00144468
CAS Number: 10004-44-1
EINECS: 233-000-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR