L(+)-Diethyl tartrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
(+)-Diethyl L-tartrate, (R,R)-(+)-Tartaric acid diethyl ester, (2R,3R)-(+)-Diethyl tartrate, Diethyl L-tartrate, L(+)-Tartaric acid diethyl ester, (2R,3R)-Diethyl 2,3-dihydroxysuccinate, (R,R)-(+)-Diethyl tartrate
132355-100G
132355-500G
BT132355-100GEA
44
USD
BT132355-100G
BT132355-500G
L(+)-Diethyl tartrate ≥98%
L(+)-Diethyl tartrate
CAS: 87-91-2; EC No: 201-783-3; MDL No: MFCD00009143
Liquid; Molecular Formula: C8H14O6 ; MW: 206.19
Boiling Point: 280°; Flash point: 93°C (199°F)
Density (g/mL): 1.204; Refractive Index: 1.446
; Optical Rotation: [α]20/D +8.5°, neat
- Optical rotation: [α]20/D +8.5°, neat
- Refractive index: 1.446
- Liquid
Formula:
C₈H₁₄O₆ Boiling Pt: 280 °C Density: 1.204 g/mL Flash Pt: 93 °C (199 °F) |
MDL Number:
MFCD00009143 CAS Number: 87-91-2 EINECS: 201-783-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...