Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Dicyclohexylphosphino-2’,6’-dimethoxybiphenyl, SPhos, Dicyclohexyl(2',6'-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine, 2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl, 2-Dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl

132350-1G 132350-25G 132350-5G
BT132350-1GEA 82.6 USD
BT132350-1G BT132350-25G BT132350-5G
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine ≥98%
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine

CAS: 657408-07-6 Solid; Molecular Formula: C26H35O2P; MW: 410.53 Melting Point: 164-166° Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₂₆H₃₅O₂P
MW: 410.53 g/mol
Melting Pt: 164…166 °C
MDL Number: MFCD05861611
CAS Number: 657408-07-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR