N-Acetyl-β-D-glucosamine ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine
132340-10G
132340-50G
BT132340-10GEA
25.57
USD
BT132340-10G
BT132340-50G
N-Acetyl-β-D-glucosamine ≥98%
N-Acetyl-β-D-glucosamine
CAS: 7512-17-6; EC No: 231-368-2; MDL No: MFCD00136044
Powder; Molecular Formula: C8H15NO6; MW: 221.21
Melting Point: 211° (decomposes)
Hygroscopic
- Hygroscopic
- Powder
Formula:
C₈H₁₅NO₆ Melting Pt: 211 °C (dec.) |
MDL Number:
MFCD00136044 CAS Number: 7512-17-6 EINECS: 231-368-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...