1,4-Bis(diphenylphosphino)butane ≥98%
Supplier: BeanTown Chemical
Synonyms:
DPPB, 1,4-Bis(diphenylphosphino)butane, Butylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)butane, Butane-1,3-diylbis(diphenylphosphine)
132275-1G
132275-25G
132275-5G
BT132275-1GEA
18
USD
BT132275-1G
BT132275-25G
BT132275-5G
1,4-Bis(diphenylphosphino)butane ≥98%
1,4-Bis(diphenylphosphino)butane
CAS: 7688-25-7; EC No: 231-698-7; MDL No: MFCD00003051
Crystalline; Linear Formula: (C6H5)2P(CH2)4P(C6H5)2; Molecular Formula: C18H28P2; MW: 426.47
Melting Point: 132-136°
- Crystalline
Formula:
C₂₈H₂₈P₂ Melting Pt: 132…136 °C |
MDL Number:
MFCD00003051 CAS Number: 7688-25-7 EINECS: 231-698-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...