Decyltrimethylammonium bromide ≥98%

Supplier: BeanTown Chemical

Synonyms: (1-Decyl)trimethylammonium bromide, N,N,N-Trimethyldecan-1-aminium bromide

132265-10G 132265-50G
BT132265-10GEA 44 USD
BT132265-10G BT132265-50G
Decyltrimethylammonium bromide ≥98%
Decyltrimethylammonium bromide

CAS: 2082-84-0; EC No: 218-219-7; MDL No: MFCD00041973; RTECS: BP6352000 Powder; Linear Formula: CH3(CH2)9N(Br)(CH3)3; Molecular Formula: C13H30BrN; MW: 280.29 Hygroscopic


  • RTECS: BP6352000
  • Hygroscopic
  • Powder

Formula: C₁₃H₃₀BrN
MW: 280.30 g/mol
MDL Number: MFCD00041973
CAS Number: 2082-84-0
EINECS: 218-219-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR