2-(4-Fluorophenyl)ethanol ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Fluorophenethyl alcohol

132260-25G 132260-5G
BT132260-25GEA 227 USD
BT132260-25G BT132260-5G
2-(4-Fluorophenyl)ethanol ≥97%
2-(4-Fluorophenyl)ethanol

CAS: 7589-27-7; EC No: 231-499-5; MDL No: MFCD00002898 Liquid; Molecular Formula: C8H9FO; MW: 140.15 Flash point: 86°C (187°F) Density (g/mL): 1.121; Refractive Index: 1.507


  • Refractive index: 1.507
  • Liquid

Formula: C₈H₉FO
MW: 140.16 g/mol
Density: 1.121 g/mL
Flash Pt: 86 °C (187 °F)
MDL Number: MFCD00002898
CAS Number: 7589-27-7
EINECS: 231-499-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR