(S)-(+)-Hydratropic acid ≥97%

Supplier: BeanTown Chemical

132245-1G 132245-250MG
BT132245-1GEA 235 USD
BT132245-1G BT132245-250MG
(S)-(+)-Hydratropic acid ≥97%
(S)-(+)-Hydratropic acid

CAS: 7782-24-3; MDL No: MFCD00063139 Crystalline; Molecular Formula: C9H10O2 ; MW: 150.17 Melting Point: 29-30°; Boiling Point: 115°/1 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.1; Refractive Index: 1.522; Optical Rotation: [α]20/D +72°, c = 1.6 in chloroform


  • Optical rotation: [α]20/D +72°, c = 1.6 in chloroform
  • Refractive index: 1.522
  • Crystalline

Formula: C₉H₁₀O₂
Boiling Pt: 115 °C/1 mmHg
Melting Pt: 29…30 °C
Density: 1.1 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00063139
CAS Number: 7782-24-3

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR