Chromium ≥99.995%, chunks 2-15 mm

Supplier: BeanTown Chemical

132235-10G 132235-50G
BT132235-10GEA 99 USD
BT132235-10G BT132235-50G
Chromium ≥99.995%, chunks 2-15 mm
Chromium

CAS: 7440-47-3; EC No: 231-157-5; MDL No: MFCD00010944; RTECS: GB4200000 Solid; Molecular Formula: Cr; MW: 52.00 Melting Point: 1857°; Boiling Point: 2672° Density (g/mL): 7.14


  • RTECS: GB4200000
  • Solid

Formula: Cr
MW: 52
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.14 g/mL
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR