Gallium nitride ≥99.99%
Supplier: BeanTown Chemical
Synonyms:
Gallium nitride, Gallium(III) nitride
132230-10G
132230-50G
BT132230-10GEA
165
USD
BT132230-10G
BT132230-50G
Gallium nitride ≥99.99%
Gallium nitride
CAS: 25617-97-4; EC No: 247-129-0; MDL No: MFCD00016108
Powder; Molecular Formula: GaN; MW: 83.73
Melting Point: 800°
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
GaN Melting Pt: 800 °C |
MDL Number:
MFCD00016108 CAS Number: 25617-97-4 EINECS: 247-129-0 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...