(±)-α-Bromo-γ-butyrolactone ≥97%
Supplier: BeanTown Chemical
CAS: 5061-21-2; EC No: 225-764-4; MDL No: MFCD00005387
Liquid; Molecular Formula: C4H5BrO2 ; MW: 164.99
Boiling Point: 138°/6 mmHg; Flash point:
- Refractive index: 1.508
- Moisture Sensitive
- Liquid
Formula:
C₄H₅BrO₂ Boiling Pt: 138 °C/6 mmHg Density: 1.786 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00005387 CAS Number: 5061-21-2 EINECS: 225-764-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...