1,1'-Bis(diphenylphosphino)ferrocene ≥97%

Supplier: BeanTown Chemical

Synonyms: DPPF, 1,1-Bis(diphenylphosphino)ferrocene

132200-1G 132200-25G 132200-5G
BT132200-1GEA 38 USD
BT132200-1G BT132200-25G BT132200-5G
1,1'-Bis(diphenylphosphino)ferrocene ≥97%
1,1'-Bis(diphenylphosphino)ferrocene

CAS: 12150-46-8; EC No: 430-420-3; MDL No: MFCD00001422 Crystalline/Powder; Molecular Formula: C34H28FeP2; MW: 554.38 Melting Point: 181-182° (decomposes) Air Sensitive


  • Air Sensitive
  • Crystalline/Powder

Formula: C₃₄H₂₈FeP₂
MW: 554.39 g/mol
Melting Pt: 181…182 °C (dec.)
MDL Number: MFCD00001422
CAS Number: 12150-46-8
EINECS: 430-420-3

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR