4-(trans-4-Propylcyclohexyl)phenyl trans-4-(trans-4-propylcyclohexyl)cyclohexanecarboxylate ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-(4-n-Propylcyclohexyl)phenyl-4'-n-propylbicyclohexyl-4-carboxylate, (Trans,trans)-4'-propyl[1,1'-Bicyclohexyl]-4-carboxyllic acid-4-(trans-4-propylcyclohexyl)phenyl ester, 4-(trans-4-Propylcyclohexyl)phenyl trans-4-(trans-4-propylcyclohexyl)cyclohexanecarboxylate

132195-1G 132195-5G
BT132195-1GEA 73 USD
BT132195-1G BT132195-5G
4-(trans-4-Propylcyclohexyl)phenyl trans-4-(trans-4-propylcyclohexyl)cyclohexanecarboxylate ≥97%
4-(trans-4-Propylcyclohexyl)phenyl trans-4-(trans-4-propylcyclohexyl)cyclohexanecarboxylate

CAS: 88038-92-0; MDL No: MFCD00673705 Solid; Molecular Formula: C31H48O2; MW: 452.72 Melting Point: 202-204°


  • Solid

Formula: C₃₁H₄₈O₂
MW: 452.723 g/mol
Melting Pt: 202…204 °C
MDL Number: MFCD00673705
CAS Number: 88038-92-0

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR