Aluminum silicate dihydrate, powder
Supplier: BeanTown Chemical
132190-100G
132190-500G
BT132190-100GEA
30
USD
BT132190-100G
BT132190-500G
Aluminum silicate dihydrate, powder
Aluminum silicate dihydrate
CAS: 1332-58-7; MDL No: MFCD11045913; RTECS: GF1670500
Powder; Molecular Formula: Al2O3·2SiO2·2H2O; MW: 258.16
Melting Point: 1750°
Density (g/mL): 2.53; Refractive Index: 1.54
- RTECS: GF1670500
- Refractive Index: 1.54
Formula:
Al₂O₃·2SiO·2H₂O Melting Pt: 1750 °C Density: 2.53 g/mL |
MDL Number:
MFCD00467010 CAS Number: 1332-58-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...