Palladium(II) trifluoroacetate ≥97% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Palladium(II) 2,2,2-trifluoroacetate
132180-1G
132180-5G
BT132180-1GEA
113
USD
BT132180-1G
BT132180-5G
Palladium(II) trifluoroacetate ≥97% (trace metals basis)
Palladium(II) trifluoroacetate
CAS: 42196-31-6; MDL No: MFCD00013204
Powder; Linear Formula: Pd(OOCCF3)2; MW: 332.44
Melting Point: 220° (decomposes)
Hygroscopic
- Hygroscopic
- Powder
Formula:
C₄F₆O₄Pd MW: 332.45 g/mol Melting Pt: 220 °C (dec.) |
MDL Number:
MFCD00013204 CAS Number: 42196-31-6 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...