3-Methoxyphenylacetone ≥97%
Supplier: BeanTown Chemical
Synonyms:
1-(3-Methoxyphenyl)propan-2-one
132125-1G
132125-5G
BT132125-1GEA
39
USD
BT132125-1G
BT132125-5G
3-Methoxyphenylacetone ≥97%
3-Methoxyphenylacetone
CAS: 3027-13-2; EC No: 221-191-9; MDL No: MFCD00008771
Liquid; Molecular Formula: C10H12O2; MW: 164.20
Boiling Point: 258-260°; Flash point:
- Refractive index: 1.525
- Liquid
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Boiling Pt: 258…260 °C Density: 1.911 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00008771 CAS Number: 3027-13-2 EINECS: 221-191-9 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...